Formamid

Lieferant: G Bioscienes
Gefahr

Synonyms: Carbamaldehyde

RC-120 RC-119
RC-120EA 417 CHF
RC-120 RC-119
Formamid
Formamid
Formel: HCONH₂
MW: 45.04 g/mol
Siedepunkt: 210
Melting Pt: 2
Dichte: 1.1334 g/cm³ (20 °C)
Flash-Pt: 79
Lager Temperatur: Kühlschrank
MDL Number: MFCD00007941
CAS-Nummer: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR