Anti-Zika Envelope Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
8669
PRSI8669EA 637 CHF
PRSI8669
Anti-Zika Envelope Rabbit Polyclonal Antibody
Antikörper
Zika Envelope antibody can be stored at 4 to 730 °C for three months and –20 to +730 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.

Type: Primary
Antigen: Zika Envelope
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zika
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR