Anti-SPT1 Rabbit Polyclonal Antibody
Lieferant: ProSci Inc.
6303
PRSI6303EA 506 CHF
PRSI6303
Anti-SPT1 Rabbit Polyclonal Antibody
Antikörper
SPT1 Antibody: Serine palmitoyltransferase, which consists of two different subunits, is the key enzyme in sphingolipid biosynthesis. It converts L-serine and palmitoyl-CoA to 3-oxosphinganine with pyridoxal 5'-phosphate as a cofactor. SPT1 is the long chain base subunit 1 of mammalian serine palmitoyltransferase. SPT1 is not catalytically active but is necessary for the stabilization of the SPT2 subunit and anchoring the holoenzyme to the cytosolic face of the endoplasmic reticulum. Missense mutations in this gene have been identified in patients with hereditary sensory neuropathy type 1 (More Product Information