Anti-SMG5 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
60-498
PRSI60-498EA 722 CHF
PRSI60-498
Anti-SMG5 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: SMG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 912-941
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR