Anti-ARG1 Mouse Monoclonal Antibody [clone: T1ARG-1]
Lieferant: ProSci Inc.
33-951
PRSI33-951EA 899 CHF
PRSI33-951
Anti-ARG1 Mouse Monoclonal Antibody [clone: T1ARG-1]
Antikörper
Arginase catalyses the hydrolysis of arginine to ornithine and urea. At least two isoforms of mammalian arginase exist (types I and II) which differ in their tissue distribution, subcellular localization, immunologic crossreactivity and physiologic function. The type I isoform is a cytosolic enzyme and expressed predominantly in the liver as a component of the urea cycle. Inherited deficiency of this enzyme results in argininemia, an autosomal recessive disorder characterised by hyperammonemia. Two transcript variants encoding different isoforms have been found for this gene. [RefSeq].