Anti-ODC1 Mouse Monoclonal Antibody [clone: ORN255-1]
Lieferant: ProSci Inc.
33-884
PRSI33-884EA 899 CHF
PRSI33-884
Anti-ODC1 Mouse Monoclonal Antibody [clone: ORN255-1]
Antikörper
Ornithine Decarboxylase (ODC) is the rate-limiting enzyme of the polyamine biosynthesis pathway which catalyses ornithine to putrescine. The activity level for the enzyme varies in response to growth-promoting stimuli and exhibits a high turnover rate in comparison to other mammalian proteins. Originally localized to both chromosomes 2 and 7, the gene encoding this enzyme has been determined to be located on 2p25, with a pseudogene located on 7q31-qter. Multiple alternatively spliced transcript variants encoding distinct isoforms have been identified. [RefSeq].
Flow Cytometry: 0,5-1 µg/More Product Information