Anti-MPI Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
22-861
PRSI22-861EA 488 CHF
PRSI22-861
Anti-MPI Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR