Anti-IKK alpha Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
2025
PRSI2025EA 506 CHF
PRSI2025
Anti-IKK alpha Rabbit Polyclonal Antibody
Antikörper
IKK alpha Antibody: Nuclear factor kappa B (NF-kappa B) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa B mediates the expression of a great variety of genes in response to extracellular stimuli including IL-1, TNFa, and bacteria product LPS. NF-kappa B is associated with I kappa B proteins in the cell cytoplasm, which inhibit NF-kappa B activity. The long-sought I kappa B kinase (IKK), which phosphorylates I kappa B, and mediates I kappa B degradation and NF-kappa B activation, was recentlyMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol IKKA
Environmentally Preferable
Antigen-Name IKK alpha
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp IgG
Western blot Yes
ELISA Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Immunogen IKK alpha antibody was raised against a peptide corresponding to amino acids 716 to 734 of human IKK alpha, which differs from corresponding murine sequence by four amino acids.
Reinigung Affinity chromatography purified via peptide column
Lagerungspuffer PBS containing 0.02% sodium azide
Validierte Anwendungen E, WB, ICC, IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR