Anti-AMG-301 Recombinant Antibody

Lieferant: ProSci Inc.
11-008
PRSI11-008EA 761 CHF
PRSI11-008
Anti-AMG-301 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4-kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR