Anti-AMG 557 Recombinant Antibody

Lieferant: ProSci Inc.
10-762
PRSI10-762EA 761 CHF
PRSI10-762
Anti-AMG 557 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2-kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR