Thorium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PTHN-100 VHG-PTHN-50 VHG-PTHN-500 VHG-TTHN-100 VHG-TTHN-500
PROOVHG-PTHN-100EA 111 CHF
PROOVHG-PTHN-100 PROOVHG-PTHN-50 PROOVHG-PTHN-500 PROOVHG-TTHN-100 PROOVHG-TTHN-500
Thorium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standards Kationischer Standards Thorium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Zertifizierungen: Traceable to NIST, SRM.
Order Now
TECHNISCHE DATEN
Environmentally Preferable