Aluminium standard solution, for ICP, in 5% HCl, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PALH-100 VHG-TALH-100 VHG-TALH-500 VHG-PALH-500
PROOVHG-PALH-100EA 99.5 CHF
PROOVHG-PALH-100 PROOVHG-TALH-100 PROOVHG-TALH-500 PROOVHG-PALH-500
Aluminium standard solution, for ICP, in 5% HCl, VHG
Standards Kationischer Standards Aluminium-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Zertifizierungen: Traceable to NIST, SRM.