Nitrite ion chromatography standard, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-INO2-100 VHG-INO2-500
PROOVHG-INO2-100EA 111 CHF
PROOVHG-INO2-100 PROOVHG-INO2-500
Nitrite ion chromatography standard, VHG
Standards Anionische Standards Nitrit-Standards
This solution is intended for use as a certified reference material (CRM) calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Zertifizierungen: Traceable to NIST and SRM.
Order Now
TECHNISCHE DATEN
Environmentally Preferable