Triethylamin

Lieferant: LGC Standards PROMOCHEM
Gefahr

Synonyms: (Diethylamino)ethane, TEA

MM0477.07
PROOMM0477.07EA 553 CHF
PROOMM0477.07
Triethylamin
Triethylamin
Formel: (C₂H₅)₃N
Siedepunkt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Dichte: 0.729 g/cm³ (20 °C)
Flash-Pt: –11 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009051
CAS-Nummer: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR