Estron

Lieferant: LGC Standards PROMOCHEM

MM0282.00
PROOMM0282.00EA 0 CHF
PROOMM0282.00
Estron
Estron
Formel: C₁₈H₂₂O₂
MDL Number: MFCD00003620
CAS-Nummer: 53-16-7
EINECS: 200-164-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR