Oxytetracyclin Hydrochlorid

Lieferant: LGC Standards PROMOCHEM

ICRS0374
PROOICRS0374EA 433 CHF
PROOICRS0374
Oxytetracyclin Hydrochlorid
Oxytetracyclin Hydrochlorid
Formel: C₂₂H₂₅ClN₂O₉
MW: 496.90 g/mol
MDL Number: MFCD00135815
CAS-Nummer: 2058-46-0
EINECS: 218-161-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR