Lidocain

Lieferant: LGC Standards PROMOCHEM

ICRS0337
PROOICRS0337EA 433 CHF
PROOICRS0337
Lidocain
Lidocain
Formel: C₁₄H₂₂N₂O
MW: 234.34 g/mol
MDL Number: MFCD00026733
CAS-Nummer: 137-58-6
EINECS: 205-302-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR