Riboflavin-5'-phosphat Natriumsalz

Lieferant: LGC Standards PROMOCHEM

Synonyms: Vitamin B2 phosphate sodium salt, Riboflavin 5'-Monophosphate sodium salt, Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogen phosphate, Flavin mononucleotide, Lactoflavin phosphate sodium salt, Riboflavin phosphate sodium

BP459
PROOBP459EA 488 CHF
PROOBP459
Riboflavin-5'-phosphat Natriumsalz
Riboflavin-5'-phosphat Natriumsalz
Formel: C₁₇H₂₀N₄NaO₉P
MDL Number: MFCD00065362
CAS-Nummer: 130-40-5
EINECS: 204-988-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR