DL-Lithiumlactat

Lieferant: LGC Standards PROMOCHEM

Synonyms: DL-Lactic acid lithium salt, (±)-2-Hydroxypropanoic acid lithium salt, Lithium 2-hydroxypropanoate, (±)-2-Hydroxypropionic acid lithium salt, (±)-Lactic acid lithium salt

BP433
PROOBP433EA 570 CHF
PROOBP433
DL-Lithiumlactat
DL-Lithiumlactat
Formel: H₃CCH(OH)COOLi
Melting Pt: 300 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00065399
CAS-Nummer: 867-55-0
EINECS: 212-761-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR