DNA- und siRNA-Transfektionsreagenz, jetPRIME®
Lieferant: Polyplus-transfection
101000001 101000046 101000027 101000015
PPLU101000001EA 5310 CHF
PPLU101000001 PPLU101000046 PPLU101000027 PPLU101000015
DNA- und siRNA-Transfektionsreagenz, jetPRIME®
Transfektionsreagenzien
The jetPRIME® DNA and/or siRNA transfection reagent is a powerful and versatile reagent designed to ensure high DNA transfection efficiency and excellent gene silencing in a variety of adherent cells.
  • High DNA transfection efficiency
  • Uses low amounts of nucleic acid
  • Superior cell viability
  • One reagent for DNA and/or siRNA transfection
  • Cost effective
jetPRIME® is ideal for co-transfection of DNA and siRNA. jetPRIME® is gentle to cells since it requires low amounts of reagent and nucleic acid during transfection. Effective and non toxic DNA and siRNA delivery is essential for reliable scientific results. Free of charge samples are available upon request. Please contact your local Avantor sales office for further details.
Since 1st of September 2021 Polyplus-Tranfection changed product part numbers. Products are remaining the same and the change is only related to the product part number. New product part numbers have symbol 'New'. For duplicated products with symbol 'telephone' please be informed that it’s the same product but with previous part number which we will only offer till the end of stock.
Zertifizierungen: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Lieferumfang: Supplied with an optimised sterile buffer which must be used to ensure successful transfection experiments. The concentrated buffer provided with 114-75C must be diluted 1:5 in sterile H₂O just prior to use. 1,5 ml is sufficient to perform ~375 transfections in 6-well plates.
Order Now
TECHNISCHE DATEN
Environmentally Preferable
DNA Ja
DNA und siRNA Ja
siRNA Ja
Virus production Ja