Plum iBox
Lieferant: Plum Safety
4921 4922 4920 4931 4923 4900
PLUM4921EA 113 CHF
PLUM4921 PLUM4922 PLUM4920 PLUM4931 PLUM4923 PLUM4900
Plum iBox
Erste-Hilfe-Ausstattung Augenduschen
The new Plum iBox is balancing key features in a modern design and is built in a strong and durable material. See-through window lets users keep track of expiry dates immediately.
  • Clearly visible eyewash bottles
  • Quick and efficient overview of expiry dates and product condition
  • Grab from the wall bracket and quickly bring to injured person
  • Spacious design allows access to bottles fast and easily
  • Easily opens – mounted on the wall or standing on the floor
  • Strong and durable material (EPP)
Release Plum iBox from the wall and quickly bring the eyewash toMore Product Information