Affinity chromatography media, HisPur™ Ni-NTA Agarose
Lieferant: Thermo Fisher Scientific
88221 88222 88223
PIER88221EA 218 CHF
PIER88221 PIER88222 PIER88223
Affinity chromatography media, HisPur™ Ni-NTA Agarose
Protein-Aufbereitungs Protein Purification Resins
HisPur™ Ni-NTA resin is a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
  • High-quality, stable and resilient affinity support
  • Bind up to 60 mg of 6xHis-tagged protein per ml of resin
  • Purify proteins using native or denaturing conditions
  • Same batch of resin can be reused at least five times
HisPur™ Ni-NTA resin consists of beaded agarose derivatised with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilised metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.