(3-Aminopropyl)triethoxysilan

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 3-Aminopropyltriethoxysilane, 3-(Triethoxysilyl)propylamine, Geniosil GF 93, Triethoxy(3-aminopropyl)silane

80370
PIER80370EA 222 CHF
PIER80370
(3-Aminopropyl)triethoxysilan
(3-Aminopropyl)triethoxysilan
Formel: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MW: 221,37 g/mol
Siedepunkt: 217 °C (1013 hPa)
Melting Pt: < –70 °C
Dichte: 0,95 g/cm³ (20 °C)
Flash-Pt: 104 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008207
CAS-Nummer: 919-30-2
EINECS: 213-048-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR