Blocking buffer, concentrate, Pierce™ Clear Milk
Lieferant: Thermo Fisher Scientific
37587
PIER37587EA 309 CHF
PIER37587
Blocking buffer, concentrate, Pierce™ Clear Milk
Puffer Elektrophorese-Puffer Blockier-Puffer
This clear milk blocking buffer is a long shelf life, high performance replacement for homemade milk blocking buffers in Western blotting applications.
  • Kit stable for 1 year stored at 4 °C
  • Convenient and saves storage space
  • Easy to use with ready-to-dilute solution
It is a proprietary pre-formulated milk solution for blocking excess non specific binding sites, reducing background in Western blotting applications and diluting antibodies when used with nitrocellulose and PVDF membranes. The solution provides lower background, enhanced sensitivity, extended shelf life and reproducible results compared to homemade buffers based on dissolved non fat dry milk. For Research Use Only. Not for use in diagnostic procedures.