Pierce™ BupH™ Wash Buffers, Dry Blend Pack
Lieferant: Thermo Fisher Scientific
28372 28382 28374 28384 28376 28379 28380 28390
PIER28372EA 346 CHF
PIER28372 PIER28382 PIER28374 PIER28384 PIER28376 PIER28379 PIER28380 PIER28390
Pierce™ BupH™ Wash Buffers, Dry Blend Pack
Puffer Elektrophorese-Puffer Waschpuffer
BupH packs are pre-blended and pre-measured dry mixtures of buffers that are easy to prepare and used in protein purification and common laboratory techniques.
  • Packs are pouches of dry-blend powder that are each sufficient to make 500 ml of buffer
  • Eliminates weighing time and pH adjustments
  • Every pack will yield the same, consistent buffer
  • Eliminates concerns about long-term stability of stock solutions
BupH™ Phosphate-Buffered Saline Packs are pouches of dry-blend powder that make sodium phosphate buffered saline (PBS) for use in crosslinking and labeling reactions requiring amine-free conditions. Each BupH dry-blend pack makes ready-to-use PBS with 0,1 M sodium phosphate, 0,15 M NaCl buffer at pH 7,2 when dissolved in 500 ml of water.
BupH MES-Buffered Saline Packs are pouches of dry-blended powder that make optimized amine-free and carboxyl-free buffer. When dissolved in 500 ml of water, the buffer contains 0,1 M 2-(N-morpholino) ethanesulfonic acid (MES), 0,9% sodium chloride at pH 4,7 for carbodiimide (EDC) crosslinking and conjugation reactions.
BupH Carbonate-Bicarbonate Buffer Packs are pouches of dry-blended powder that are each sufficient to make 500 ml of alkaline (pH 9,4) sodium-carbonate (carb-bicarb) buffer. This buffer is used for coating or binding capture-antibodies and other proteins onto polystyrene microplates for ELISA and other plate assays. When dissolved in 500 ml of water, each pack makes 0,2 M carbonate-bicarbonate at pH 9,4.
BupH Tris-Buffered Saline Packs are pouches of dry-blend powder that make Tris-NaCl buffered saline (TBS) for diluents and wash buffers in ELISA, western blotting, and general immunoassay procedures. Each pack makes ready-to-use TBS with 25 mm Tris, 0,15 M NaCl at pH 7,2 when dissolved in 500 ml of water.
BupH Borate Buffer Packs are pouches of dry-blended powder that are each sufficient to make borate buffer for use in protein modification procedures requiring amine-free conditions. When dissolved in 500 ml of water, each pack makes 50 mM borate at pH 8,5.
BupH Tris-Glycine Transfer Buffer Packs are pouches of dry-blend powder that make 500 ml of standard transfer buffer for wet or semi-dry electrophoretic protein transfer from gel to blotting membrane. Each pack makes 25 mm Tris, 192 mM glycine at pH 8 when dissolved in 400 ml of water and 100 ml methanol.
BupH Modified Dulbecco's PBS Packs are pouches of dry-blend powder that make physiological sodium- and potassium-phosphate buffered saline (D-PBS) for ELISA and western blot diluents and wash buffers. Each pack makes 8 mm sodium phosphate, 2 mm potassium phosphate, 0,14 M NaCl, 10 mm KCl modified Dulbecco's PBS buffer at pH 7,4 when dissolved in 500 ml of water.
Bitte beachten For research use only. Not for use in diagnostic procedures.
Order Now
TECHNISCHE DATEN
Environmentally Preferable