Affinity purification resin, immobilised p-Aminophenyl phosphoryl choline gel, Pierce™

Lieferant: Thermo Fisher Scientific

20307
PIER20307EA 725 CHF
PIER20307
Affinity purification resin, immobilised p-Aminophenyl phosphoryl choline gel, Pierce™
Protein-Aufbereitungs Protein Purification Resins

Pierce™ Immobilised p-Aminophenyl Phosphoryl Choline Gel enables purification of C-reactive protein (CRP) from plasma, ascites and other biological fluids.


  • Format: 50% resin slurry in 0,02% sodium azide
  • Binding condition: Tris or borate-buffered saline (pH 8 to 8,5) with 1 to 2 mM calcium chloride
  • Elution condition: Tris or borate-buffered saline (pH 8 to 8,5) with 2 mM EDTA
  • Reusable: resin can be regenerated and reused several times

Order Now

TECHNISCHE DATEN

Ligand p-aminophenyl phosphoryl choline
Environmentally Preferable
Matrix Crosslinked 6% beaded agarose
Bindungskapazität >3 mg of human C-reactive protein/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR