Pharmaceutical secondary standard, Phenytoin, certified reference material, Supelco®
Lieferant: Merck
PHR1139-1G
PHR1139-1GEA 148 CHF
PHR1139-1G
Pharmaceutical secondary standard, Phenytoin, certified reference material, Supelco®
Standards Organische Standards
Traceable to PhEur P1290000 and to USP 1535008.
Reduces incidence of grand mal seizures; appears to stabilise excitable membranes perhaps through effects on Na⁺, K⁺, and Ca²⁺ channels.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Zertifizierungen: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=104b3cf0247f4c11b55609f20766ae8903f2f0e3&vid=104b3cf0247f4c11b55609f20766ae8903f2f0e3&prid=web&clid=SAPDAM