Scandium Standard Solution, 1,000 μg/ml, 2% HNO₃
Lieferant: PerkinElmer
N9303751
PERCN9303751EA 141 CHF
PERCN9303751
Scandium Standard Solution, 1,000 μg/ml, 2% HNO₃
Standards Kationischer Standards Scandium-Standards
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppb level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Bitte beachten These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).