Electrodeless discharge lamps
Lieferant: PerkinElmer
N3050605 N3050611 N3050615 N3050620 N3050630 N3050634 N3050655 N3050657 N3050664 N3050670 N3050672 N3050675 N3050680 N3050683 N3050691
PERCN3050605EA 1760 CHF
PERCN3050605 PERCN3050611 PERCN3050615 PERCN3050620 PERCN3050630 PERCN3050634 PERCN3050655 PERCN3050657 PERCN3050664 PERCN3050670 PERCN3050672 PERCN3050675 PERCN3050680 PERCN3050683 PERCN3050691
Electrodeless discharge lamps
Beleuchtung Hohlkathodenlampen
These electrodeless discharge lamps are preferred for certain volatile elements. They offer better precision and lower detection limits for analyses that are 'noisy' due to weak hollow cathode emission. PerkinElmer system 2 EDLs consist of the element or a salt of the element sealed in a quartz bulb containing an inert gas atmosphere. When an RF field of sufficient power is applied, the inert gas is ionised and the coupled energy vaporizes the element and excites the atoms inside the bulb, resulting in the emission of the characteristic spectrum.
  • Interchangeable lamp sleeves
  • Dual-
More Product Information