Ceramic D-Torch™
Lieferant: PerkinElmer
N0777840
PERCN0777840EA 1460 CHF
PERCN0777840
Ceramic D-Torch™
Analysatoren Elementar-Analysatoren
The ceramic outer tube is of particular benefit for the analysis of wear metals in engine oils, where quartz outer tubes often suffer from short lifetime.
  • Analysis of high-matrix solutions – Fusion samples, samples with high-dissolved solids, samples with high-acid content
  • Demountable quart or ceramic outer tubes
Also beneficial for Si determinations, where quartz outer tubes may produce high background signals.
Can cause early devitrification or sample deposition when using quartz.