Mixing Systems, SpinBag
Lieferant: PARKER HANNIFIN
PHSBU010LGI PHSBU050LGI
PARKPHSBU010LGIEA 0 CHF
PARKPHSBU010LGI PARKPHSBU050LGI
Mixing Systems, SpinBag
Rührer Rührwerke
Parker's compact SpinBag Mixing System offers the advantages of single-use in a bench top mixing solution.
  • Constructed of fully validated DuraPure C93 film offering added seal strength, low gas permeability and optical clarity which can be heated to 60 °C
  • Stores flat and ideal for preparation of cell culture media, buffers, bioreactor feeds, and suspension/re-suspension applications
  • Easily integrated with filters, sensors and pumping systems
  • Available with standard configurations, or a variety of ports and tubing options in non-irradiated or gamma
More Product Information