Poly-L-lysine hydrobromide

Lieferant: SIGMA ALDRICH MICROSCOPY

P8920-100ML P8920-500ML
P8920-100MLEA 262 CHF
P8920-100ML P8920-500ML
Poly-L-lysine hydrobromide
Proteine und Peptide

Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.


Poly-L-lysine polymers can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations. Lower molecular weight poly-L-lysine (30,000-70,000) is less viscuous in solution, but higher molecular weight versions provide more attachment sites per molecule.


This product is supplied as a 0.10% (w/v) solution in water. Coating slides with this solution should be followed with a 5 minute incubation and drying the slides at room temperature. Coated slides will be stable for one year if protected from dust.

Order Now

TECHNISCHE DATEN

Protein/Peptid-Name Poly-L-lysine hydrobromide
Environmentally Preferable
Spezies
Ursprung
CAS Nr. 25988-63-0
MDL No. MFCD00237230
Konzentration 0.1 % w/v
Zusammensetzung Solution in water
Molecular weight 150000 - 300000
Molecular formula (C₆H₁₃N₂OBr)n
Lagerbedingungen Ambient
M-Clarity Qualitätsstufe MQ200

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR