Catecholviolett Indicator, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyms: Pyrocatechol Violet

MQ200

P7884-5G P7884-10G
P7884-5GEA 135 CHF
P7884-5G P7884-10G
Catecholviolett Indicator, Sigma-Aldrich®
Catecholviolett

Pyrocatechol Violet (PV) is a sulfone phthalein dye. It chelates metal ions. The metal-PV complex is bluish-violet in colour.


  • Complexometric indicator
  • Pyrocatechol Violet has been used in the formation of dye-metal complexes
  • It has also been used to determine Cu²⁺ chelating activity

Formel: C₁₉H₁₄O₇S
MW: 386.38 g/mol
Melting Pt: 185 °C (Zers., Lit.)
Lager Temperatur: Room Temperature
MDL Number: MFCD00005868
CAS-Nummer: 115-41-3
EINECS: 204-088-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06602426615c86a927ad7e90a851130ceed633f2&vid=d5f7e466702b450a34f0bd56c256049c825b0de4&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Form) Powder
1H NMR Spectrum Conforms to Structure
Solubility (Colour), c = 10 mg/10 ml; Water Faint Brown to Brown and Faint Red to Dark Red
Suitability, Suitable for use as an indicator for EDTA titrations Suitable
Suitability, Suitable for use as a pH indicator Suitable

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR