Natriumthiosulfat Pentahydrat Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

P121-33EA 51.4 CHF
P121-33
Natriumthiosulfat Pentahydrat Ph. Eur., Macron Fine Chemicals™
Natriumthiosulfat Pentahydrat
Formel: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48
MDL Number: MFCD00149186
CAS-Nummer: 10102-17-7
Merck Index: 12,08844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR