Magnesiumoxid Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyms: Magnesia usta

P119-33EA 69.6 CHF
P119-33
Magnesiumoxid Ph. Eur., Macron Fine Chemicals™
Magnesiumoxid
Formel: MgO
MDL Number: MFCD00011109
CAS-Nummer: 1309-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR