Anti-CD22 Mouse Monoclonal Antibody [clone: OTI4C3]

Lieferant: OriGene
TA506412
ORIGTA506412EA 723 CHF
ORIGTA506412
Anti-CD22 Mouse Monoclonal Antibody [clone: OTI4C3]
Antikörper
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150

Type: Primary
Antigen: CD22 (CD22 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR