Anti-PACSIN3 Mouse Monoclonal Antibody [clone: OTI4C5]

Lieferant: OriGene
TA502240
ORIGTA502240EA 723 CHF
ORIGTA502240
Anti-PACSIN3 Mouse Monoclonal Antibody [clone: OTI4C5]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100

Type: Primary
Antigen: PACSIN3 (protein kinase C and casein kinase substrate in neurons 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR