Anti-CCR5 Armenian Hamster Polyclonal Antibody [clone: HM-CCR5 (7A4)]

Lieferant: OriGene
TA320231
ORIGTA320231EA 410 CHF
ORIGTA320231
Anti-CCR5 Armenian Hamster Polyclonal Antibody [clone: HM-CCR5 (7A4)]
Antikörper
Type: Primary
Antigen: CCR5
Clonality: Polyclonal
Clone: HM-CCR5 (7A4)
Conjugation: Unconjugated
Epitope:
Host: Armenian Hamster
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR