Anti-BRCA1 (Ab-2) (MS13) Mouse Monoclonal Antibody [clone: MS13]

Lieferant: Merck Millipore (Oncogene)
OP93-100
OP93-100EA 1070 CHF
OP93-100
Anti-BRCA1 (Ab-2) (MS13) Mouse Monoclonal Antibody [clone: MS13]
Antikörper
Type: Primary
Antigen: BRCA1 (Ab-2) (MS13)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR