Anti-MDM2 (Ab-5) (4B2C1.11) Mouse Monoclonal Antibody [clone: 4B2C1.11]

Lieferant: Merck Millipore (Oncogene)
OP145-100
OP145-100EA 1100 CHF
OP145-100
Anti-MDM2 (Ab-5) (4B2C1.11) Mouse Monoclonal Antibody [clone: 4B2C1.11]
Antikörper
Type: Primary
Antigen: MDM2 (Ab-5) (4B2C1.11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR