GMF syringe filters, Target®
Lieferant: THERMO NATIONAL SCIENTIFIC
F2500-18 F2500-19 F2500-20
NSCAF2500-18EA 568 CHF
NSCAF2500-18 NSCAF2500-19 NSCAF2500-20
GMF syringe filters, Target®
Filter Spritzenvorsatzfilter
The Target® syringe filters are ideal for general purpose laboratory use. Target2® GMF (Glass MicroFiber) syringe filters are used for dissolution testing. They are also well-suited for general filtration applications.
  • Low extractable membranes and housings ensure that no filter-borne contaminants are added to your sample
  • Performance-tested under actual HPLC conditions
  • 30 mm products are pressure rated to 120 psi (8,3 bar)
  • Able to filter solutions at temperatures up to 100 °C
Housings are manufactured from solvent-resistant, low-extractable PP resinsMore Product Information