Anti-Mouse Monoclonal Antibody [clone: rMT1/410] (FITC)

Lieferant: Novus Biologicals
NBP3-08824F
NOVUNBP3-08824FEA 894 CHF
NOVUNBP3-08824F
Anti-Mouse Monoclonal Antibody [clone: rMT1/410] (FITC)
Antikörper
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: rMT1/410
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR