Anti-Fractin Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP2-29504
NOVUNBP2-29504EA 0 CHF
NOVUNBP2-29504
Anti-Fractin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Fractin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

TECHNISCHE DATEN

Environmentally Preferable
Antigen-Name Fractin
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Immunogen Synthetic peptide corresponding to amino acid residues from the C terminal region of the 32 kDa actin fragment
Lagerungspuffer Neat whole antisera
Lagertemperatur Store at −20 °C. Avoid freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR