Anti-HMGB3 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP2-24645
NOVUNBP2-24645EA 505 CHF
NOVUNBP2-24645
Anti-HMGB3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HMGB3 (high mobility group box 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR