Anti-LC3I Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Novus Biologicals
NBP1-78964H
NOVUNBP1-78964HEA 840 CHF
NOVUNBP1-78964H
Anti-LC3I Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: LC3I
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR