Anti-STK33 Rabbit Polyclonal Antibody (DyLight® 550)

Lieferant: Novus Biologicals
NBP1-54156R
NOVUNBP1-54156REA 777 CHF
NOVUNBP1-54156R
Anti-STK33 Rabbit Polyclonal Antibody (DyLight® 550)
Antikörper
The STK33 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to STK33. This antibody reacts with human, mouse. The STK33 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: STK33
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

TECHNISCHE DATEN

Antigen-Symbol STK33
Environmentally Preferable
Antigen-Name Serine/threonine Kinase 33
Antigen-Synonyme EC 2.7.11
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation DyLight® 550
Reaktivität Human
Wirt Rabbit
Gen-ID 65975
Isotyp
ImmunFluoreszenz Yes
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR