Anti-BIRC3 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Novus Biologicals
NBP1-27972F
NOVUNBP1-27972FEA 777 CHF
NOVUNBP1-27972F
Anti-BIRC3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: BIRC3 (baculoviral IAP repeat containing 3)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR