Anti-WRN Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-471
NOVUNB100-471EA 860 CHF
NOVUNB100-471
Anti-WRN Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal WRN Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: WRN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR