Apparatus for the determination of aromatic oils by steam distillation according to DAB 7
Lieferant: NEUBERT VOLUME GLASSWAERE
2410-10-100
NEUB2410-10-100EA 940 CHF
NEUB2410-10-100
Apparatus for the determination of aromatic oils by steam distillation according to DAB 7
Extraktoren Flüssig-Flüssig-Extraktoren
Steam distillation apparatus designed to extract and determine the resulting volume aromatic oils. Apparatus has graduated receiver for organic matter for 10 ml volume in 0,1 ml increments.