Tanks, zylindrisch, besonders strapazierfähige Ausführung, mit Deckel, Nalgene®
Lieferant: Thermo Fisher Scientific
11100-0005 11100-0007 11100-0010 11100-0015 11100-0030 11100-0055 11100-0080 11100-0100 11100-0150 11100-0200
NALG11100-0005EA 330 CHF
NALG11100-0005 NALG11100-0007 NALG11100-0010 NALG11100-0015 NALG11100-0030 NALG11100-0055 NALG11100-0080 NALG11100-0100 NALG11100-0150 NALG11100-0200
Tanks, zylindrisch, besonders strapazierfähige Ausführung, mit Deckel, Nalgene®
Tanks Tanks, zylindrisch
Heavy-duty cylindrical LLDPE tanks are offered in sizes ranging from 19 to 757 liters and include covers to signficantly reduce evaporation and contamination.
  • Good temperature tolerance
  • With factory-installed spigots
  • Not autoclavable
Zubehörinformationen: Flexible tank liners: Coex polyethylene film tank liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene® cylindrical plastic tanks
Designed to fit Nalgene® cylindrical tanks from 5 gal (19 liters) to 200 gal (757 liters)
Gamma-irradiated,More Product Information